- Genetic evidence that Nkx2.2 acts primarily downstream of Neurog3 in pancreatic endocrine lineage development.
Genetic evidence that Nkx2.2 acts primarily downstream of Neurog3 in pancreatic endocrine lineage development.
eLife (2017-01-11)
Angela J Churchill, Giselle Dominguez Gutiérrez, Ruth A Singer, David S Lorberbaum, Kevin A Fischer, Lori Sussel
PMID28071588
RÉSUMÉ
Many pancreatic transcription factors that are essential for islet cell differentiation have been well characterized; however, because they are often expressed in several different cell populations, their functional hierarchy remains unclear. To parse out the spatiotemporal regulation of islet cell differentiation, we used a
MATÉRIAUX
Référence du produit
Marque
Description du produit
Sigma-Aldrich
Anti-NKX2-2 antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution