Merck
  • Matrix-assisted laser desorption/ionization mass spectrometry of acidic glycoconjugates facilitated by the use of spermine as a co-matrix.
Matrix-assisted laser desorption/ionization mass spectrometry of acidic glycoconjugates facilitated by the use of spermine as a co-matrix.
Journal of the American Society for Mass Spectrometry (1998-12-03)
Y Mechref, M V Novotny
RÉSUMÉ
Negative-ion matrix-assisted laser desorption/ionization mass spectra of sialyated glycoconjugates were acquired employing 2,5-dihydroxybenzoic acid (DHB) in conjunction with spermine as a co-matrix. The addition of spermine to DHB permitted an improved crystal formation as well as a higher analyte solubility. Moreover, DHB/spermine appears to minimize alkali adduct formation, thus allowing the sample analysis without desalting. The combined matrix permitted the analysis of complex sialylated and sialylated/fucosylated structures down to the femotomole range. The ability to use such a matrix also facilitates determination of the sialic acid linkages (in combination with a specific enzyme cleavage). The matrix also appears suitable for studies on gangliosides.
MATÉRIAUX
Référence du produit
Marque
Description du produit
Sigma-Aldrich
Spermine, ≥99.0% (GC)