Merck
  • Synthesis and Pharmacological Activity of a New Series of 1-(1H-Indol-4-yloxy)-3-(2-(2-methoxyphenoxy)ethylamino)propan-2-ol Analogs.
Synthesis and Pharmacological Activity of a New Series of 1-(1H-Indol-4-yloxy)-3-(2-(2-methoxyphenoxy)ethylamino)propan-2-ol Analogs.
Archiv der Pharmazie (2016-02-09)
Marek Bednarski, Monika Otto, Magdalena Dudek, Marcin Kołaczkowski, Adam Bucki, Agata Siwek, Grażyna Groszek, Elżbieta Maziarz, Piotr Wilk, Jacek Sapa
RÉSUMÉ
β-Adrenergic receptor antagonists are important therapeutics for the treatment of cardiovascular disorders. In the group of β-blockers, much attention is being paid to the third-generation drugs that possess important ancillary properties besides inhibiting β-adrenoceptors. Vasodilating activity of these drugs is produced through different mechanisms, such as nitric oxide (NO) release, β2 -agonistic action, α1 -blockade, antioxidant action, and Ca(2+) entry blockade. Here, a study on evaluation of the cardiovascular activity of five new compounds is presented. Compound 3a is a methyl and four of the tested compounds (3b-e) are dimethoxy derivatives of 1-(1H-indol-4-yloxy)-3-(2-(2-methoxyphenoxy)ethylamino)propan-2-ol. The obtained results confirmed that the methyl and dimethoxy derivatives of 1-(1H-indol-4-yloxy)-3-(2-(2-methoxyphenoxy)ethylamino)propan-2-ol and their enantiomers possess α1 - and β1 -adrenolytic activities and that the antiarrhythmic and hypotensive effects of the tested compounds are related to their adrenolytic properties.
MATÉRIAUX
Référence du produit
Marque
Description du produit
Sigma-Aldrich
4-Hydroxyindole, 99%