Merck
  • Piperazine derivatives of benzimidazole as potential anthelmintics. Part 1: Synthesis and activity of methyl-5-(4-substituted piperazin-1-yl)benzimidazole-2-carbamates.
Piperazine derivatives of benzimidazole as potential anthelmintics. Part 1: Synthesis and activity of methyl-5-(4-substituted piperazin-1-yl)benzimidazole-2-carbamates.
Die Pharmazie (1989-09-01)
R M Sánchez-Alonso, E Raviña, L Santana, G García-Mera, M Sanmartín, P Baltar
RÉSUMÉ
A series of 5-(4-substituted piperazin-1-yl)-2-nitroanilines (4) and 5-(4-substituted piperazin-1-yl)benzimidazole-2-carbamates (6) has been synthesized starting from 5-chloro-2-nitroaniline (3) and N-monosubstituted piperazines. Catalytic reduction of 4 with Pd/C followed by treatment with 1,3-dicarbomethoxy-S-methylisothiourea yielded the corresponding methyl-5-(4-substituted piperazin-1-yl)benzimidazole-2-carbamates (6) which were for anthelmintic activity against experimental infections of Trichinella spiralis.
MATÉRIAUX
Référence du produit
Marque
Description du produit
Sigma-Aldrich
5-Chloro-2-nitroaniline, 97%