Merck
  • Health and ecological risk-based characterization of soil and sediment contamination in shipyard with long-term use of DDT-containing antifouling paint.
Health and ecological risk-based characterization of soil and sediment contamination in shipyard with long-term use of DDT-containing antifouling paint.
The Science of the total environment (2013-03-19)
Guanlin Guo, Chao Zhang, Guanglong Wu, Qiong Ding, Shijie Wang, Fasheng Li
RÉSUMÉ
Dichlorodiphenyltrichloroethane (DDT) was a frequently occurring type of persistent organic environmental pollutant in China and DDT-containing antifouling paint could be the main contributor of DDT to shipyards and fishing harbors. A field survey was conducted in a shipyard in southern China to investigate the content and distribution of DDT in soil and sediments. Human health and screening-level ecological risk assessments were conducted for DDT contamination in soil and sediments and the results indicated that total DDT in all samples tested exceeded present advisory safe limits. Analysis of the composition and distribution implicated DDT-containing antifouling paint used for ship maintenance as an important source of DDT. Individual and cumulative health risks for residents exceeded the extra lifetime cancer risks of 10(-6) and 10(-5), mainly from exposure to soil, ingestion and dermal contact. DDT in sediments is associated with a high level of toxicity for the benthic community when >99% of samples exceed the threshold concentration likely to be responsible for effects and severe effects. Further risk control for DDT is required to ensure safety for human health, the benthic community and the environment.
MATÉRIAUX
Référence du produit
Marque
Description du produit
Supelco
4,4'-DDT solution, certified reference material, 5000 μg/mL in methanol
Supelco
4,4'-DDT, PESTANAL®, analytical standard