Merck
Preparation and properties of electrophoretic microcapsules for electronic paper.
Journal of microencapsulation (2007-04-19)
J K Song, H J Choi, I Chin
RÉSUMÉ
This paper shows two types of microcapsules used for electrophoretic display. One is prepared by in-situ polymerization which is based on urea, melamine and formaldehyde and another by complex coacervation, which is composed of gelatin and gum Arabic. Microcapsules attract interests of many research groups for longer lifetime of electrophoretic display by reducing agglomerization or lateral movements of nanoparticles. The gelatin microcapsules were more attractive in providing more uniform microcapsule coverage on electrodes due to their flexibility as compared to the melamine-urea microcapsules. The properties of microcapsules were characterized by FTIR, OM, SEM and TGA. Migration of nanoparticles in the two types of microcapsules was also observed when an electric field was applied.
MATÉRIAUX
Référence du produit
Marque
Description du produit
Sigma-Aldrich
Poly(ethylene-alt-maleic anhydride), average Mw 100,000-500,000, powder