Merck
The histone demethylase KDM2B activates FAK and PI3K that control tumor cell motility.
Cancer biology & therapy (2020-03-17)
Nefeli Zacharopoulou, Galatea Kallergi, Saad Alkahtani, Anna Tsapara, Saud Alarifi, Evi Schmid, Basma Sukkar, Sotirios Kampranis, Florian Lang, Christos Stournaras
RÉSUMÉ
Recent studies revealed that the histone demethylase KDM2B regulates the epithelial markers E-Cadherin and ZO-1, the RhoA/B/C-small-GTPases and actin cytoskeleton organization, in DU-145 prostate- and HCT-116 colon-tumor cells. Here we addressed the role of KDM2B in the activation of Focal Adhesion Kinase (FAK)-signaling and its involvement in regulating tumor cell motility. We used RT-PCR for gene transcriptional analysis, Western blotting for the assessment of protein expression and activity and wound-healing assay for the study of cell migration. KDM2B overexpression or silencing controls the activity of FAK in DU-145 prostate- and HCT-116 colon-tumor cells without affecting gene transcription and protein expression of this kinase. Upon KDM2B overexpression in DU-145 cells, significantly enhanced migration was observed, which was abolished in cells pretreated by the specific phosphoinositide-3 kinase (PI3 K) inhibitor LY294002, implying involvement of FAK/PI3 K signaling in the migration process. In line with this, the p85-PI3 K-subunit was downregulated upon knockdown of KDM2B in DU-145 cells, while the opposite effect became evident in KDM2B-overexpressing cells. These results revealed a novel functional role of KDM2B in regulating the activation of the FAK/PI3 K signaling in prostate cancer cells that participates in the control of cell motility.
MATÉRIAUX
Référence du produit
Marque
Description du produit
Sigma-Aldrich
Anticorps anti-JHDM1B, from rabbit, purified by affinity chromatography