Key Documents
TMO140654
Nunc® Cell Culture Inserts in 12-well Nunclon Delta surface plate
cell culture treated, polycarbonate, sterile, pore size 3.0 μm, case of 48 ea (supplied in 4 x 12 well plates)
Synonyme(s) :
multidish membrane insert, treated membrane insert
About This Item
Code UNSPSC :
41122102
Nomenclature NACRES :
NB.11
Matériaux
polycarbonate
Stérilité
sterile
Conditionnement
case of 48 ea (supplied in 4 x 12 well plates)
Fabricant/nom de marque
Nunc 140654
Dimension de pores
3.0 μm pore size
Type de liaison
cell culture treated
Description générale
Cultivate most cell types without matrix coating when using Thermo Scientific Nunc Polycarbonate Membrane Inserts in Multidishes.
Informations légales
Nunc is a registered trademark of Thermo Fisher Scientific or its subsidiaries