Key Documents
Z186724
Atomizer bulb
useful for priming liquid-transfer lines
Synonyme(s) :
priming bulk
About This Item
Code UNSPSC :
41121500
Nomenclature NACRES :
NB.01
Matériaux
rubber bulb
Conditionnement
pack of 1 ea
Fabricant/nom de marque
205A
Diamètre
53 mm
Volume
75 mL
Description générale
Rubber bulb with hard-rubber valves. Useful for priming liquid-transfer lines.