Key Documents
Z177024
LDPE wash bottles
Wide-mouth, capacity 500 mL
Synonyme(s) :
bottle lab, laboratory bottles, wash bottle
About This Item
Code UNSPSC :
41103209
Nomenclature NACRES :
NB.17
Matériaux
low-density polyethylene bottle
natural bottle
polypropylene cap
wide-mouth bottle
Joint
Joints threaded neck (53)
Caractéristiques
label no label
vented
with cap
Conditionnement
pack of 6 ea
Fabricant/nom de marque
Bel-Art F11620-0500
Capacité
500 mL
Description générale
Consist of one-piece cap and nozzle plus delivery tube. Deliver single drops or a needle-sharp stream. Tip may be cut to increase delivery amounts. Polyallomer cap.