Key Documents
Z148512
Sigma-Aldrich
Aldrich® corrosive lecture-bottle gas regulator
delivery pressure 1-50 psig, CGA Inlet size 180
Synonyme(s) :
Corrosive Gas Regulator, Lecture-Bottle Regulator
About This Item
Code UNSPSC :
41121800
Nomenclature NACRES :
NB.51
delivery pressure measuring range
1-50 psig
Taille de l'entrée CGA
180
Description générale
  • Single-stage
  • CGA inlet connection, needle valve outlet with ¼ in. NPTM connection
  • Max. inlet pressure: 0-4000 psig
  • Temp. range: -40 to 140°F
  • 316 Stainless Steel body 1 ½ in. gauge face, PTFE seals and inner friction sleeves
  • NPTF bonnet-vent (1/8 in.) connection for piping regulator to fume hood in case of diaphragm failure; order bonnet-vent tubing connectors separately
  • Use with "T"-purge valves (listed separately) when regulating corrosive gases to minimize gas hydrolysis and corrosion of equipment
  • Use for high-purity applications
Lecture-bottle gas regulators include a PTFE sealing washer. Order spare PTFE sealing washers separately.
Autres remarques
Informations légales
Aldrich is a registered trademark of Sigma-Aldrich Co. LLC