Key Documents
Z146145
Sigma-Aldrich
Plier-type decapper
O.D. 11 mm
About This Item
Code UNSPSC :
41121800
Nomenclature NACRES :
NB.11
D.E.
11 mm
Description générale
Removes aluminum seals from serum bottles and vials.