Z131008
Nalgene® beaker with handle
capacity 3,000 mL
Synonyme(s) :
HDPE beaker, heavy wall beaker
About This Item
Produits recommandés
Matériaux
high-density polyethylene
Caractéristiques
graduations
Conditionnement
pack of 1 ea
Fabricant/nom de marque
Nalgene 1220-2500
Capacité
3,000 mL
Description générale
Heavy-wall, HDPE, rectangular handle. Graduated in oz and mL. Suitable for handling acids and alkalis.
Autres remarques
Browse all plastic beakers
Informations légales
Nalgene is a registered trademark of Thermo Fisher Scientific or its subsidiaries