Skip to Content
MilliporeSigma
Skip To
SYNC212440
Synthware Friedrichs condenser
top joint: ST/NS 24/40, bottom joint: ST/NS 24/40
Synonym(s):
Synthware C212440, glass Friedrichs condenser
Sign In to View Organizational & Contract Pricing.
Select a Size
Change View
Size/SKUAvailabilityPrice
1 ea
Available to ship TODAYfromMILWAUKEE
$177.00
About This Item
NACRES:
NB.51
UNSPSC Code:
41121800
$177.00
Available to ship TODAYDetails
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist
joint
bottom joint: ST/NS24/40, joint: ST/NS top24/40
packaging
pkg of 1 ea
manufacturer/tradename
Synthware C212440
General description
With a joint at 15º angle.
Legal Information
Synthware is a trademark of Kemtech America Inc.
Compare Similar Items
View Full Comparison
Show Differences
1 of 1
This Item
SYNC214550SYNC243445SYNC232440
packaging
pkg of 1 ea
packaging
pkg of 1 ea
packaging
pkg of 1 ea
packaging
pack of 1 ea
joint
bottom joint: ST/NS24/40, joint: ST/NS top24/40
joint
bottom joint: ST/NS45/50, joint: ST/NS top24/40
joint
joint: ST/NS34/45
joint
bottom joint: ST/NS24/40
manufacturer/tradename
Synthware C212440
manufacturer/tradename
Synthware C214550
manufacturer/tradename
Synthware C243445
manufacturer/tradename
C232440
Still not finding the right product?
Explore all of our products under Synthware Friedrichs condenser
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Lot/Batch Number
If you need assistance, please contact Customer Support
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Visit the Document Library