Key Documents
BR23809
BRAND® compact automatic SILBERBRAND burette, without support/clamp
volume 10 mL, with LDPE bottle and PVC filling tube
Synonyme(s) :
Automatic Burettes, Burettes, Compact Automatic Burettes, SILBERBRAND
About This Item
Code UNSPSC :
41121808
Nomenclature NACRES :
NB.43
Matériaux
AR-Glass
Fabricant/nom de marque
BRAND 23809
Plage de mesure du volume
10 mL, accuracy: ±0.05 mL
Volume
10 mL
Description générale
The Modular Automatic Burette Concept! Fast to dismantle and easy to clean. Easy to repair - all individual components replaceable! Boro 3.3, detachable PTFE stopcock. Error limits according to class B, DIN EN ISO 385. Calibrated to deliver (TD, Ex).
Informations légales
BRAND is a registered trademark of BRAND GMBH + CO KG