Merck
HomeSmall Molecule HPLCHPLC Analysis of Polyphenols in Nero d'Avola Red Wine on Discovery® HS C18 (UV 280 nm)
HPLC Analysis of Polyphenols in Nero d'Avola Red Wine on Discovery® HS C18 (UV 280 nm)
HPLC Analysis of Polyphenols in Nero d'Avola Red Wine
Conditions
sample/matrixNero d'Avola wine sample filtered prior injection through a 0.45 µm nylon membrane
columnDiscovery HS C18, 15 cm x 2.1 mm I.D., 3 µm particles (569255-U)
column temp.30 °C
mobile phase[A] water, pH 3 with formic acid; [B] acetonitrile, pH 3 with formic acid
flow rate0.2 mL/min
injection5 µL
detectorUV, 280 nm
Description
CategoriesAnalytical Chromatography, Clinical, Food & Beverage Analysis, phenols, Flavors, Fragrances, Beverage Testing, flavors/fragrances, Beer, Wine, and Spirits, Wine
Featured IndustryFood and Beverages
Legal InformationDiscovery is a registered trademark of Sigma-Aldrich Co. LLC
suitabilityapplication for HPLC
Materials
Sorry, an unexpected error has occurred
Network error: Failed to fetch