HPLC Analysis of Miconazole Enantiomers on Astec® CYCLOBOND I 2000 RSP
CONDITIONS
column
CYCLOBOND I 2000 RSP, 25 cm x 4.6 mm I.D., 5 μm particles (20324AST)
mobile phase
[A] 1% triethylamine acetate, pH 4.1; [B] acetonitrile; (80:20, A:B)
flow rate
2 mL/min
column temp.
ambient
detector
UV, 230 nm
injection
5 μL
sample
5 mg/mL in methanol
Description
Legal Information
Astec is a registered trademark of Merck KGaA, Darmstadt, Germany
null