US EPA Method 8015: GC Analysis of Nonhalogenated Volatiles on SLB®-5ms
![US EPA Method 8015: GC Analysis of Nonhalogenated Volatiles on SLB®-5ms suitable for GC](/deepweb/assets/sigmaaldrich/product/images/391/687/b2963536-577d-4862-8ead-3a18017d8cc2/800/b2963536-577d-4862-8ead-3a18017d8cc2.jpg)
Materials
analytical column
Product No.
Description
Pricing
CONDITIONS
column
SLB-5ms, 30 m x 0.25 mm I.D., 1.0 μm (28476-U)
oven
50 °C (3 min), 15 °C/min to 200 °C
inj. temp.
150 °C
detector
FID, 250 °C
carrier gas
helium, 1.3 mL/min., constant
injection
0.5 μL 50:1 split
liner
2 mm I.D., FocusLiner™ inlet liner
sample
each analyte at 2000 μg/mL in methanol
Description
Legal Information
FocusLiner is a trademark of SGE Analytical Science Pty Ltd.
SLB is a registered trademark of Merck KGaA, Darmstadt, Germany