Merck
  • Low-Polarization Lithium-Oxygen Battery Using [DEME][TFSI] Ionic Liquid Electrolyte.
Low-Polarization Lithium-Oxygen Battery Using [DEME][TFSI] Ionic Liquid Electrolyte.
ChemSusChem (2017-09-30)
Ulderico Ulissi, Giuseppe Antonio Elia, Sangsik Jeong, Franziska Mueller, Jakub Reiter, Nikolaos Tsiouvaras, Yang-Kook Sun, Bruno Scrosati, Stefano Passerini, Jusef Hassoun
ABSTRACT
The room-temperature molten salt mixture of N,N-diethyl-N-(2-methoxyethyl)-N-methylammonium bis(trifluoromethanesulfonyl) imide ([DEME][TFSI]) and lithium bis(trifluoromethanesulfonyl)imide (LiTFSI) salt is herein reported as electrolyte for application in Li-O
Sigma-Aldrich
N,N-Diethylmethylamine, 97%
Sigma-Aldrich
2-Bromoethyl methyl ether