Merck
  • Pharmacological modulation of AMPA receptor phosphorylation by dopamine and muscarinic receptor agents in the rat medial prefrontal cortex.
Pharmacological modulation of AMPA receptor phosphorylation by dopamine and muscarinic receptor agents in the rat medial prefrontal cortex.
European journal of pharmacology (2017-12-16)
Bing Xue, Li-Min Mao, Dao-Zhong Jin, John Q Wang
ABSTRACT
Two key transmitters in the medial prefrontal cortex (mPFC), dopamine and acetylcholine, are believed to interact with each other to modulate local glutamatergic transmission, although molecular mechanisms underlying their crosstalk are poorly understood. Here we investigated effects of pharmacological manipulations of dopamine and muscarinic receptors on phosphorylation of α-amino-3-hydroxy-5-methyl-4-isoxazolepropionic acid (AMPA) receptors in the adult rat mPFC in vivo. We found that an agonist selective for G
Sigma-Aldrich
VU0152100, ≥98% (HPLC)
Sigma-Aldrich
S-(−)-Eticlopride hydrochloride, ≥98% (HPLC), powder
Sigma-Aldrich
(−)-Quinpirole hydrochloride, ≥98% (HPLC), solid