Merck
  • Alcohol-abuse drug disulfiram targets cancer via p97 segregase adaptor NPL4.
Alcohol-abuse drug disulfiram targets cancer via p97 segregase adaptor NPL4.
Nature (2017-12-07)
Zdenek Skrott, Martin Mistrik, Klaus Kaae Andersen, Søren Friis, Dusana Majera, Jan Gursky, Tomas Ozdian, Jirina Bartkova, Zsofia Turi, Pavel Moudry, Marianne Kraus, Martina Michalova, Jana Vaclavkova, Petr Dzubak, Ivo Vrobel, Pavla Pouckova, Jindrich Sedlacek, Andrea Miklovicova, Anne Kutt, Jing Li, Jana Mattova, Christoph Driessen, Q Ping Dou, Jørgen Olsen, Marian Hajduch, Boris Cvek, Raymond J Deshaies, Jiri Bartek
ABSTRACT
Cancer incidence is rising and this global challenge is further exacerbated by tumour resistance to available medicines. A promising approach to meet the need for improved cancer treatment is drug repurposing. Here we highlight the potential for repurposing disulfiram (also known by the trade name Antabuse), an old alcohol-aversion drug that has been shown to be effective against diverse cancer types in preclinical studies. Our nationwide epidemiological study reveals that patients who continuously used disulfiram have a lower risk of death from cancer compared to those who stopped using the drug at their diagnosis. Moreover, we identify the ditiocarb-copper complex as the metabolite of disulfiram that is responsible for its anti-cancer effects, and provide methods to detect preferential accumulation of the complex in tumours and candidate biomarkers to analyse its effect on cells and tissues. Finally, our functional and biophysical analyses reveal the molecular target of disulfiram's tumour-suppressing effects as NPL4, an adaptor of p97 (also known as VCP) segregase, which is essential for the turnover of proteins involved in multiple regulatory and stress-response pathways in cells.
Sigma-Aldrich
cis-11-Methyl-2-dodecenoic acid, ≥90.0% (HPLC)
Sigma-Aldrich
1,10-Phenanthroline, ≥99%
Sigma-Aldrich
Copper(II) D-gluconate, 98%
Sigma-Aldrich
DBeQ, ≥98% (HPLC)
Sigma-Aldrich
Anti-Histone H2A (acidic patch) Antibody, serum, Upstate®
Sigma-Aldrich
Anti-ATF4 Antibody, serum, from rabbit
Sigma-Aldrich
CuET, ≥97% (HPLC)
Sigma-Aldrich
Z-Leu-Leu-Glu-7-amido-4-methylcoumarin, ≥95%, solid
Sigma-Aldrich
Z-Leu-Leu-Leu-al, ≥90% (HPLC)