Merck
  • Functionalized polyacrylamide as an acetylcholinesterase-inspired biomimetic device for electrochemical sensing of organophosphorus pesticides.
Functionalized polyacrylamide as an acetylcholinesterase-inspired biomimetic device for electrochemical sensing of organophosphorus pesticides.
Biosensors & bioelectronics (2017-09-25)
Livia F Sgobbi, Sergio A S Machado
ABSTRACT
A plethora of publications has continuously reported electrochemical biosensors for detection of pesticides. However, those devices rarely accomplish commercial application due to technical issues associated with the lack of stability and high cost of the biological recognition element (enzyme). Alternatively, the biomimetic catalysts have arisen as a candidate for application in electrochemical biosensors to overcome the enzymatic drawbacks, combining low cost scalable materials with superior stability. Herein, for the first time, we propose a biomimetic biosensor for organophosphorus pesticide detection employing a functionalized polyacrylamide, polyhydroxamicalkanoate (PHA), which mimics the performance of the acetylcholinesterase (AChE) enzyme. The PHA bears functional groups inserted along its backbone chain working as active sites. Thereby, PHA was immobilized on screen printed electrodes (SPE) through a blend formation with poly(ethylene glycol) methyl ether (mPEG) to prevent its leaching out from the surface. Under optimum conditions, the biomimetic sensor was employed for the amperometric detection of paraoxon-ethyl, fenitrothion and chlorpyrifos ranging from 1.0 and 10.0μmolL
Sigma-Aldrich
6-Phosphonohexanoic acid, 97%
Sigma-Aldrich
Acetylthiocholine chloride, ≥99% (TLC), powder