Merck
  • TLR4-Mediated Inflammation Promotes KSHV-Induced Cellular Transformation and Tumorigenesis by Activating the STAT3 Pathway.
TLR4-Mediated Inflammation Promotes KSHV-Induced Cellular Transformation and Tumorigenesis by Activating the STAT3 Pathway.
Cancer research (2017-10-21)
Marion Gruffaz, Karthik Vasan, Brandon Tan, Suzane Ramos da Silva, Shou-Jiang Gao
ABSTRACT
Toll-like receptors (TLR) are conserved immune sensors mediating antimicrobial and antitumoral responses, but recent evidence implicates them in promoting carcinogenesis in certain cancers. Kaposi sarcoma is caused by infection of Kaposi sarcoma-associated herpesvirus (KSHV) and is characterized by uncontrolled neoangiogenesis and inflammation. Here, we show that TLR4 is upregulated in KSHV-infected spindle tumor cells in human Kaposi sarcoma lesions. In a model of KSHV-induced cellular transformation, KSHV upregulated expression of TLR4, its adaptor MyD88, and coreceptors CD14 and MD2. KSHV induction of TLR4 was mediated by multiple viral miRNAs. Importantly, the TLR4 pathway was activated constitutively in KSHV-transformed cells, resulting in chronic induction of IL6, IL1β, and IL18. Accordingly, IL6 mediated constitutive activation of the STAT3 pathway, an essential event for uncontrolled cellular proliferation and transformation. TLR4 stimulation with lipopolysaccharides or live bacteria enhanced tumorigenesis while TLR4 antagonist CLI095 inhibited it. These results highlight an essential role of the TLR4 pathway and chronic inflammation in KSHV-induced tumorigenesis, which helps explain why HIV-infected patients, who frequently suffer from opportunistic bacterial infections and metabolic complications, frequently develop Kaposi sarcoma. Cancer Res; 77(24); 7094-108. ©2017 AACR.
Sigma-Aldrich
SC144 hydrochloride, ≥98% (HPLC)
Sigma-Aldrich
LookOut® Mycoplasma PCR Detection Kit, Optimized for use with JumpStart Taq DNA Polymerase, D9307.
Sigma-Aldrich
Anti-IκBα antibody produced in rabbit, IgG fraction of antiserum, buffered aqueous solution
Avanti
Lactosyl(β) Sphingosine (d18:1), Avanti Research - A Croda Brand 860542P, powder
Sigma-Aldrich
TRI Reagent®, For processing tissues, cells cultured in monolayer or cell pellets