Merck
  • Favorable outcome of experimental islet xenotransplantation without immunosuppression in a nonhuman primate model of diabetes.
Favorable outcome of experimental islet xenotransplantation without immunosuppression in a nonhuman primate model of diabetes.
Proceedings of the National Academy of Sciences of the United States of America (2017-10-29)
Barbara Ludwig, Stefan Ludwig, Anja Steffen, Yvonne Knauf, Baruch Zimerman, Sophie Heinke, Susann Lehmann, Undine Schubert, Janine Schmid, Martina Bleyer, Uwe Schönmann, Clark K Colton, Ezio Bonifacio, Michele Solimena, Andreas Reichel, Andrew V Schally, Avi Rotem, Uriel Barkai, Helena Grinberg-Rashi, Franz-Josef Kaup, Yuval Avni, Peter Jones, Stefan R Bornstein
ABSTRACT
Transplantation of pancreatic islets for treating type 1 diabetes is restricted to patients with critical metabolic lability resulting from the need for immunosuppression and the shortage of donor organs. To overcome these barriers, we developed a strategy to macroencapsulate islets from different sources that allow their survival and function without immunosuppression. Here we report successful and safe transplantation of porcine islets with a bioartificial pancreas device in diabetic primates without any immune suppression. This strategy should lead to pioneering clinical trials with xenotransplantation for treatment of diabetes and, thereby, represents a previously unidentified approach to efficient cell replacement for a broad spectrum of endocrine disorders and other organ dysfunctions.
Sigma-Aldrich
Monoclonal Anti-Glucagon antibody produced in mouse, clone K79bB10, ascites fluid
Sigma-Aldrich
Anti-CD8 antibody, Rabbit monoclonal, clone SP16, recombinant, expressed in proprietary host, tissue culture supernatant
Sigma-Aldrich
Anti-CD3, T Cell antibody produced in rabbit, whole antiserum