Merck
  • Asiatic acid protects against hepatic ischemia/reperfusion injury by inactivation of Kupffer cells via PPARγ/NLRP3 inflammasome signaling pathway.
Asiatic acid protects against hepatic ischemia/reperfusion injury by inactivation of Kupffer cells via PPARγ/NLRP3 inflammasome signaling pathway.
Oncotarget (2017-11-22)
Ying Xu, Jun Yao, Chen Zou, Heng Zhang, Shouliang Zhang, Jun Liu, Gui Ma, Pengcheng Jiang, Wenbo Zhang
ABSTRACT
Hepatic ischemia/reperfusion (I/R) contributes to major complications in clinical practice affecting perioperative morbidity and mortality. Recent evidence suggests the key role of nucleotide-binding oligomerization domain-like receptor (NLR) family pyrin domain-containing 3 (NLRP3) inflammaosme activation on the pathogenesis of I/R injury. Asiatic acid (AA) is a pentacyclic triterpene derivative presented with versatile activities, including antioxidant, anti-inflammation and hepatoprotective effects. This study was designed to determine whether AA had potential hepatoprotective benefits against hepatic I/R injury, as well as to unveil the underlying mechanisms involved in the putative effects. Mice subjected to warm hepatic I/R, and Kupffer cells (KCs) or RAW264.7 cells challenged with lipopolysaccharide (LPS)/H
Sigma-Aldrich
MISSION® esiRNA, targeting human NLRP3
Sigma-Aldrich
MISSION® esiRNA, targeting human PYCARD
Sigma-Aldrich
MISSION® esiRNA, targeting human AIM2