Merck
  • The Short Isoform of BRD4 Promotes HIV-1 Latency by Engaging Repressive SWI/SNF Chromatin-Remodeling Complexes.
The Short Isoform of BRD4 Promotes HIV-1 Latency by Engaging Repressive SWI/SNF Chromatin-Remodeling Complexes.
Molecular cell (2017-08-29)
Ryan J Conrad, Parinaz Fozouni, Sean Thomas, Hendrik Sy, Qiang Zhang, Ming-Ming Zhou, Melanie Ott
ABSTRACT
BET proteins commonly activate cellular gene expression, yet inhibiting their recruitment paradoxically reactivates latent HIV-1 transcription. Here we identify the short isoform of BET family member BRD4 (BRD4S) as a corepressor of HIV-1 transcription. We found that BRD4S was enriched in chromatin fractions of latently infected T cells, and it was more rapidly displaced from chromatin upon BET inhibition than the long isoform. BET inhibition induced marked nucleosome remodeling at the latent HIV-1 promoter, which was dependent on the activity of BRG1-associated factors (BAF), an SWI/SNF chromatin-remodeling complex with known repressive functions in HIV-1 transcription. BRD4S directly bound BRG1, a catalytic subunit of BAF, via its bromodomain and extraterminal (ET) domain, and this isoform was necessary for BRG1 recruitment to latent HIV-1 chromatin. Using chromatin immunoprecipitation sequencing (ChIP-seq) combined with assay for transposase-accessible chromatin coupled to high-throughput sequencing (ATAC-seq) data, we found that the latent HIV-1 promoter phenotypically resembles endogenous long terminal repeat (LTR) sequences, pointing to a select role of BRD4S-BRG1 complexes in genomic silencing of invasive retroelements.
Product Description
Millipore
ANTI-FLAG® antibody produced in rabbit, affinity isolated antibody, buffered aqueous solution
Sigma-Aldrich
Anti-BAF180 Antibody, from rabbit
Sigma-Aldrich
Monoclonal ANTI-FLAG® M2 antibody produced in mouse, 1 mg/mL, clone M2, affinity isolated antibody, buffered aqueous solution (50% glycerol, 10 mM sodium phosphate, and 150 mM NaCl, pH 7.4)