Merck
  • MKK4 activates non-canonical NFκB signaling by promoting NFκB2-p100 processing.
MKK4 activates non-canonical NFκB signaling by promoting NFκB2-p100 processing.
Biochemical and biophysical research communications (2017-07-25)
Jeong Seon Kim, Eun Ju Kim, Hee-Sun Kim, Jonathan M Kurie, Young-Ho Ahn
ABSTRACT
The NFκB family of transcription factors is crucial for innate or adaptive immunity, inflammation, and diseases including cancer. The two NFκB signaling pathways (canonical and non-canonical) differ from each other in extracellular signals, membrane receptors, signaling adaptors, and dimer subunits. The p52 (NFκB2) subunit, which participates in the non-canonical pathway, is generated by ubiquitin-mediated processing of the p100 precursor. Here, we found that NFκB2 processing and activation were mediated by mitogen-activated protein kinase kinase-4 (MKK4) and its substrate c-Jun N-terminal kinase (JNK). In MKK4-null mouse embryonic fibroblasts (MEFs), serum- and lymphotoxin β receptor (LTβR) antibody-induced processing of p100 and nuclear translocation of p52 were found to be defective. Serum and LTβR antibody activated the MKK4-JNK signaling pathway, and SP600125, a JNK inhibitor, blocked p100 processing. Cellular senescence, one of the responses regulated by the non-canonical NFκB pathway, was observed more frequently in MKK4-null MEFs than in wildtype cells. These results suggest that the MKK4/JNK-dependent pathway regulates NFκB2 processing/activation and, through this mechanism, MKK4 and NFκB2 control cellular growth and senescence.
Sigma-Aldrich
SP600125, ≥98% (HPLC)
Sigma-Aldrich
Triton X-100, laboratory grade
Sigma-Aldrich
Z-Leu-Leu-Leu-al, ≥90% (HPLC)
Sigma-Aldrich
MISSION® esiRNA, targeting human NFKB2