Merck
  • Oligo-Fucoidan prevents IL-6 and CCL2 production and cooperates with p53 to suppress ATM signaling and tumor progression.
Oligo-Fucoidan prevents IL-6 and CCL2 production and cooperates with p53 to suppress ATM signaling and tumor progression.
Scientific reports (2017-09-21)
Li-Mei Chen, Po-Yen Liu, Yen-An Chen, Hong-Yu Tseng, Pei-Chun Shen, Pai-An Hwang, Hsin-Ling Hsu
ABSTRACT
Low-molecular-weight Fucoidan (Oligo-Fucoidan) is a sulfated polysaccharide that has a variety of biological effects and has also been shown to have beneficial health effects. However, the molecular mechanisms underlying the therapeutic effects of Oligo-Fucoidan in patients with cancer remain unclear. Using human colorectal cancer HCT116 cells with (p53
Sigma-Aldrich
Fucoidan from Fucus vesiculosus, ≥95%
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Triton X-100, laboratory grade