Merck
Evaluation of nicotine in tobacco-free-nicotine commercial products.
Drug testing and analysis (2016-12-13)
Garrett Hellinghausen, Jauh T Lee, Choyce A Weatherly, Diego A Lopez, Daniel W Armstrong
ABSTRACT
Recently, a variety of new tobacco-free-nicotine, TFN, products have been commercialized as e-liquids. Tobacco-derived nicotine contains predominantly (S)-(-)-nicotine, whereas TFN products may not. The TFN products are said to be cleaner, purer substances, devoid of toxic components that come from the tobacco extraction process. A variety of commercial tobacco and TFN products were analyzed to identify the presence and composition of each nicotine enantiomer. A rapid and effective enantiomeric separation of nicotine has been developed using a modified macrocyclic glycopeptide bonded to superficially porous particles. The enantiomeric assay can be completed in <2 min with high resolution and accuracy using high performance liquid chromatography with electrospray ionization mass spectrometry. The results of this study suggest the need for pharmacological studies of (R)-(+)-nicotine, which is present in much greater quantities in commercial TFN products compared to commercial tobacco-derived products. Such studies are required by the FDA for new enantiomeric pharmacological products. Copyright © 2016 John Wiley & Sons, Ltd.
Sigma-Aldrich
Ammonium trifluoroacetate, 98%