Merck
  • Digestibility of Glyoxal-Glycated β-Casein and β-Lactoglobulin and Distribution of Peptide-Bound Advanced Glycation End Products in Gastrointestinal Digests.
Digestibility of Glyoxal-Glycated β-Casein and β-Lactoglobulin and Distribution of Peptide-Bound Advanced Glycation End Products in Gastrointestinal Digests.
Journal of agricultural and food chemistry (2017-06-28)
Di Zhao, Lin Li, Thao T Le, Lotte Bach Larsen, Guoying Su, Yi Liang, Bing Li
ABSTRACT
This work reports the influence of glyoxal (GO)-derived glycation on the gastrointestinal enzymatic hydrolysis of β-lactoglobulin and β-casein. Reduced digestibility of glycated proteins was found in both gastric and intestinal stage. Distribution of Maillard reaction products in digests with different molecular weight ranges was investigated subsequently. The colorless and brown MRPs largely presented in the digests smaller than 20 kDa. However, the resistance of fluorescent advanced glycation end products (AGEs) to enzymatic hydrolysis gradually increased during glycation, rendering fluorescent AGEs largely present in the digests larger than 20 kDa. No free N (ε)-carboxymethyllysine (CML) was detected in digests. The relative amount of CML in digests larger than 1 kDa was higher than that of Lys, demonstrating the hindrance of CML to enzymatic hydrolysis. This study highlights the resistance of GO-derived AGEs to digestive proteases via blockage of tryptic cleavage sites or steric hindrance, which is a barrier to the absorption of dietary AGEs.
Sigma-Aldrich
Gly-Gly-Tyr-Arg, ≥97% (HPLC)