Merck
  • Cellular imaging by green luminescence of Tb(III)-doped aminomodified silica nanoparticles.
Cellular imaging by green luminescence of Tb(III)-doped aminomodified silica nanoparticles.
Materials science & engineering. C, Materials for biological applications (2017-05-10)
Svetlana V Fedorenko, Asiya R Mustafina, Alsu R Mukhametshina, Michail E Jilkin, Timur A Mukhametzyanov, Anastasiya O Solovieva, Tatiana N Pozmogova, Lidiya V Shestopalova, Michael A Shestopalov, Kirill V Kholin, Yury N Osin, Oleg G Sinyashin
ABSTRACT
The work introduces Tb(III)-centered luminescence of amino-modified silica nanoparticles doped with Tb(III) complexes for cellular imaging. For these purposes water-in-oil procedure was optimized for synthesis of 20 and 35nm luminescent nanoparticles with amino-groups embedded on the surface. The obtained results indicate an impact of the nanoparticle size in decoration, aggregation behavior and luminescent properties of the nanoparticles in protein-based buffer solutions. Formation of a protein-based corona on the nanoparticles surface was revealed through the effect of the nanoparticles on helical superstructure of BSA. This effect is evident from CD spectral data, while no any size impact on the adsorption of BSA onto aminomodified silica surface was observed. Cellular uptake of the nanoparticles studied by confocal and TEM microscopy methods indicates greater cellular uptake for the smaller nanoparticles. Cytotoxicity of the nanoparticles was found to agree well with their cellular uptake behavior, which in turn was found to be greater for the smaller nanoparticles.
Sigma-Aldrich
Terbium(III) nitrate hexahydrate, 99.999% trace metals basis
Pricing and availability is not currently available.
Sigma-Aldrich
Triton X-100, laboratory grade
Pricing and availability is not currently available.