Merck
  • Determining the effect of different cooking methods on the nutritional composition of salmon (Salmo salar) and chilean jack mackerel (Trachurus murphyi) fillets.
Determining the effect of different cooking methods on the nutritional composition of salmon (Salmo salar) and chilean jack mackerel (Trachurus murphyi) fillets.
PloS one (2017-07-08)
José M Bastías, Pamela Balladares, Sergio Acuña, Roberto Quevedo, Ociel Muñoz
ABSTRACT
The effect of four cooking methods was evaluated for proximate composition, fatty acid, calcium, iron, and zinc content in salmon and Chilean jack mackerel. The moisture content of steamed salmon decreased (64.94%) compared to the control (68.05%); a significant decrease was observed in Chilean jack mackerel in all the treatments when compared to the control (75.37%). Protein content in both salmon and Chilean jack mackerel significantly increased under the different treatments while the most significant decrease in lipids was found in oven cooking and canning for salmon and microwaving for Chilean jack mackerel. Ash concentration in both salmon and Chilean jack mackerel did not reveal any significant differences. Iron and calcium content only had significant changes in steaming while zinc did not undergo any significant changes in the different treatments. Finally, no drastic changes were observed in the fatty acid profile in both salmon and Chilean jack mackerel.
Product Description
Supelco
C4 - C24 Even Carbon Saturated FAMEs, 1000 μg/mL each component in hexane, analytical standard
Pricing and availability is not currently available.