Merck
Cancer with low cathepsin D levels is susceptible to vacuolar (H
Cancer science (2017-03-21)
Satoshi Kitazawa, Satoru Nishizawa, Hideyuki Nakagawa, Masaaki Funata, Kazuho Nishimura, Tomoyoshi Soga, Takahito Hara
ABSTRACT
Vacuolar (H
Sigma-Aldrich
MISSION® esiRNA, targeting human CTSD, RP11-295K3.1
Sigma-Aldrich
Acridine Orange hydrochloride solution, 10 mg/mL in H2O, ≥95.0% (HPLC)
Sigma-Aldrich
5-(N-Ethyl-N-isopropyl)amiloride