Merck
  • Exosomes bind to autotaxin and act as a physiological delivery mechanism to stimulate LPA receptor signalling in cells.
Exosomes bind to autotaxin and act as a physiological delivery mechanism to stimulate LPA receptor signalling in cells.
Journal of cell science (2016-08-26)
Susanna A Jethwa, Emma J Leah, Qifeng Zhang, Nicholas A Bright, David Oxley, Martin D Bootman, Simon A Rudge, Michael J O Wakelam
ABSTRACT
Autotaxin (ATX; also known as ENPP2), the lysophospholipase responsible for generating the lipid receptor agonist lysophosphatidic acid (LPA), is a secreted enzyme. Here we show that, once secreted, ATX can bind to the surface of cell-secreted exosomes. Exosome-bound ATX is catalytically active and carries generated LPA. Once bound to a cell, through specific integrin interactions, ATX releases the LPA to activate cell surface G-protein-coupled receptors of LPA; inhibition of signalling by the receptor antagonist Ki1642 suggests that these receptors are LPAR1 and LPAR3. The binding stimulates downstream signalling, including phosphorylation of AKT and mitogen-activated protein kinases, the release of intracellular stored Ca2+ and cell migration. We propose that exosomal binding of LPA-loaded ATX provides a means of efficiently delivering the lipid agonist to cell surface receptors to promote signalling. We further propose that this is a means by which ATX-LPA signalling operates physiologically.
Sigma-Aldrich
Anti-phospho-Erk1/2 (Thr202/Tyr204, Thr185/Tyr187)Antibody, recombinant clone AW39R, rabbit monoclonal, clone AW39R, Upstate®, from rabbit
Sigma-Aldrich
Anti-LAMC1 antibody produced in rabbit, Ab1, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution