Merck
  • Correlations between Structure and Near-Infrared Spectra of Saturated and Unsaturated Carboxylic Acids. Insight from Anharmonic Density Functional Theory Calculations.
Correlations between Structure and Near-Infrared Spectra of Saturated and Unsaturated Carboxylic Acids. Insight from Anharmonic Density Functional Theory Calculations.
The journal of physical chemistry. A (2017-04-18)
Justyna Grabska, Mika Ishigaki, Krzysztof B Beć, Marek J Wójcik, Yukihiro Ozaki
ABSTRACT
By near-infrared (NIR) spectroscopy and anharmonic density functional theory (DFT) calculations, we investigate five kinds of saturated and unsaturated carboxylic acids belonging to the group of short-chain fatty acids: propionic acid, butyric acid, acrylic acid, crotonic acid, and vinylacetic acid. The experimental NIR spectra of these five kinds of carboxylic acids are reproduced by quantum chemical calculations in a broad spectral region of 7500-4000 cm
Sigma-Aldrich
3-Butenoic acid, 97%
Sigma-Aldrich
Crotonic acid, 98%