Merck
  • Serum A08 C1q antibodies are associated with disease activity and prognosis in Chinese patients with lupus nephritis.
Serum A08 C1q antibodies are associated with disease activity and prognosis in Chinese patients with lupus nephritis.
Kidney international (2016-10-18)
Yun Pang, Ying Tan, Yongzhe Li, Jianchun Zhang, Yongbing Guo, Zhiling Guo, Chengying Zhang, Feng Yu, Ming-Hui Zhao
ABSTRACT
Systemic lupus erythematosus (SLE) is an autoimmune disease characterized by overproduction of numerous autoantibodies. Many studies have sought to identify such biomarkers to distinguish patients with active lupus nephritis from SLE patients without renal involvement. Because antibodies to complement C1q appear to be prevalent in patients with active lupus nephritis, we analyzed the frequency of antigenic epitopes of C1q and their clinical significance in a large multicenter study of Chinese patients. The lupus cohort consisted of 210 patients with active lupus nephritis as a discovery cohort, 130 active patients as a validation cohort along with 130 SLE patients without clinical renal involvement, and 100 healthy controls. Serum antibodies to intact C1q, the collagen-like region, the globular head region, and the new linear A08 epitope to C1q were screened by specific ELISA. The frequency of antibodies to intact C1q, the C1q-collagen-like region, and the A08 antibodies in the discovery cohort were significantly higher than that in patients without renal involvement or healthy controls. Antibodies to the globular head region were not prevalent enough for further study. The results were confirmed in the validation cohort. The area under the curve for anti-A08 antibodies was significantly greater than those for both the intact and collagen-like region antibodies to discriminate between active lupus nephritis and active SLE without clinical renal involvement. The A08 antibodies were all negative at remission. The serum A08 antibody level correlated better with disease relapse than that of antibodies to either the intact or the collagen-like region, significantly predicting renal prognosis. Thus, serum levels of A08 C1q antibodies are closely associated with disease activity and prognosis in lupus nephritis.
Sigma-Aldrich
Somatotropin from human pituitary, recombinant, expressed in E. coli, ≥95% (SDS-PAGE), lyophilized powder