Merck
  • TRPA1 Mediates Aromatase Inhibitor-Evoked Pain by the Aromatase Substrate Androstenedione.
TRPA1 Mediates Aromatase Inhibitor-Evoked Pain by the Aromatase Substrate Androstenedione.
Cancer research (2016-10-21)
Francesco De Logu, Raquel Tonello, Serena Materazzi, Romina Nassini, Camilla Fusi, Elisabetta Coppi, Simone Li Puma, Ilaria M Marone, Laura R Sadofsky, Alyn H Morice, Tommaso Susini, Alessandro Terreni, Gloriano Moneti, Mariarosaria Di Tommaso, Pierangelo Geppetti, Silvia Benemei
ABSTRACT
Aromatase inhibitors (AI) induce painful musculoskeletal symptoms (AIMSS), which are dependent upon the pain transducing receptor TRPA1. However, as the AI concentrations required to engage TRPA1 in mice are higher than those found in the plasma of patients, we hypothesized that additional factors may cooperate to induce AIMSS. Here we report that the aromatase substrate androstenedione, unique among several steroid hormones, targeted TRPA1 in peptidergic primary sensory neurons in rodent and human cells expressing the native or recombinant channel. Androstenedione dramatically lowered the concentration of letrozole required to engage TRPA1. Notably, addition of a minimal dose of androstenedione to physiologically ineffective doses of letrozole and oxidative stress byproducts produces AIMSS-like behaviors and neurogenic inflammatory responses in mice. Elevated androstenedione levels cooperated with low letrozole concentrations and inflammatory mediators were sufficient to provoke AIMSS-like behaviors. The generation of such painful conditions by small quantities of simultaneously administered TRPA1 agonists justifies previous failure to identify a precise link between AIs and AIMSS, underscoring the potential of channel antagonists to treat AIMSS. Cancer Res; 76(23); 7024-35. ©2016 AACR.
Sigma-Aldrich
4-Androstenedione, ≥98% (HPLC)
Sigma-Aldrich
Reichstein′s substance S, ≥98%