Merck
  • Targeted disruption of the voltage-dependent calcium channel alpha2/delta-1-subunit.
Targeted disruption of the voltage-dependent calcium channel alpha2/delta-1-subunit.
American journal of physiology. Heart and circulatory physiology (2009-05-12)
Geraldine A Fuller-Bicer, Gyula Varadi, Sheryl E Koch, Masakazu Ishii, Ilona Bodi, Nijiat Kadeer, James N Muth, Gabor Mikala, Natalia N Petrashevskaya, Michael A Jordan, Sui-Po Zhang, Ning Qin, Christopher M Flores, Idit Isaacsohn, Maria Varadi, Yasuo Mori, W Keith Jones, Arnold Schwartz
ABSTRACT
Cardiac L-type voltage-dependent Ca(2+) channels are heteromultimeric polypeptide complexes of alpha(1)-, alpha(2)/delta-, and beta-subunits. The alpha(2)/delta-1-subunit possesses a stereoselective, high-affinity binding site for gabapentin, widely used to treat epilepsy and postherpetic neuralgic pain as well as sleep disorders. Mutations in alpha(2)/delta-subunits of voltage-dependent Ca(2+) channels have been associated with different diseases, including epilepsy. Multiple heterologous coexpression systems have been used to study the effects of the deletion of the alpha(2)/delta-1-subunit, but attempts at a conventional knockout animal model have been ineffective. We report the development of a viable conventional knockout mouse using a construct targeting exon 2 of alpha(2)/delta-1. While the deletion of the subunit is not lethal, these animals lack high-affinity gabapentin binding sites and demonstrate a significantly decreased basal myocardial contractility and relaxation and a decreased L-type Ca(2+) current peak current amplitude. This is a novel model for studying the function of the alpha(2)/delta-1-subunit and will be of importance in the development of new pharmacological therapies.
Sigma-Aldrich
Monoclonal Anti-Dihydropyridine Receptor (α2 Subunit) antibody produced in mouse, clone 20A, ascites fluid, buffered aqueous solution
Sigma-Aldrich
Anti-Cav2.2 Antibody, Chemicon®, from rabbit
Sigma-Aldrich
Anti-Calcium Channel (α1A Subunit) (P/Q-type of Voltage-gated Ca2+ Channel) antibody produced in rabbit, affinity isolated antibody, lyophilized powder