Merck
  • Preparation of Magnetic Tubular Nanoreactors for Highly Efficient Catalysis.
Preparation of Magnetic Tubular Nanoreactors for Highly Efficient Catalysis.
Chemistry, an Asian journal (2016-04-29)
Shuliang Yang, Li Peng, Changyan Cao, Fang Wei, Jian Liu, Ya-Nan Zhu, Chang Liu, Xiaoshi Wang, Weiguo Song
ABSTRACT
We report an efficient and controllable route to prepare magnetic tubular nanoreactors composed of a mesoporous SiO
Sigma-Aldrich
3,5-Di-tert-butylcatechol, 98%
Sigma-Aldrich
4-Chlorostyrene, 97%, contains 500 ppm 4-tert-butylcatechol as inhibitor
Sigma-Aldrich
Triethylene glycol, ReagentPlus®, 99%
Sigma-Aldrich
trans-Stilbene, 96%
Sigma-Aldrich
4-Vinylanisole, 97%