Merck
Curcumin inhibits proliferation of interleukin-22-treated HaCaT cells.
International journal of clinical and experimental medicine (2015-08-27)
Yi Zhao, Jun Sun, Wei Dou, Jin-Hong Hu
ABSTRACT
The interleukin (IL)-23/IL-17A/IL-22 cytokine axis plays a critical role in the pathogenesis of psoriasis, wherein IL-22 effects on epidermal alternations by inhibiting differentiation and inducing keratinocyte proliferation. In this study, we investigated effects of curcumin on IL-22-induced proliferation in a human keratinocyte cell line (HaCaT) in vitro experiment. The HaCaT cells growth was measured by a Cell Counting Kit-8. The cyclin D1 and cyclin E was detected by real-time RT-PCR and weatern blot. The STAT3 and phosphorylation of STAT3 was tested by weatern blot also. Our results show that curcumin exhibited a significant anti-proliferation effect on HaCaT cells, even in the presence of IL-22. Since STAT3 is crucial for IL-22 signal transduction, we examined the level of phosphorylation of STAT3 in all of the experimental groups. As expected, curcumin inhibited IL-22 induced phosphorylation of STAT3; furthermore, curcumin down regulated cyclin D1 and cyclin E. We can reach a conclusion that curcumin can suppress the proliferation of keratinocytes even with IL-22 treatment. Therefore, we have confidence in future curcumin research about psoriasis treatment.
Sigma-Aldrich
IL-22 human, Animal-component free, recombinant, expressed in E. coli, ≥98% (SDS-PAGE), ≥98% (HPLC)
Sigma-Aldrich
IL-22 human, recombinant, expressed in E. coli, ≥98% (SDS-PAGE), ≥98% (HPLC), suitable for cell culture
Sigma-Aldrich
Curcumin, ≥94% (curcuminoid content), ≥80% (Curcumin)
Sigma-Aldrich
Curcumin, from Curcuma longa (Turmeric), powder