Merck
One-step protecting-group-free synthesis of azepinomycin in water.
Organic & biomolecular chemistry (2015-02-07)
Adam J Coggins, Derek A Tocher, Matthew W Powner
ABSTRACT
We report an efficient, atom economical general acid-base catalyzed one-step multi-gram synthesis of azepinomycin from commercially available compounds in water. We propose that the described pH-dependent Amadori rearrangement, which couples an amino-imidazole and simple sugar, is of importance as a potential step toward predisposed purine nucleotide synthesis at the origins of life.
Sigma-Aldrich
Glycolaldehyde dimer, crystalline, mixture of stereoisomers. Melts between 80 and 90 °C depending on stereoisomeric composition
Sigma-Aldrich
5-Amino-4-imidazolecarboxamide, 95%
Dacarbazine impurity B, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
5-Amino-4-imidazolecarboxamide hydrochloride, 98%