Merck
  • Ni(2+) affects dopamine uptake which limits suitability as inhibitor of T-type voltage-gated Ca(2+) channels.
Ni(2+) affects dopamine uptake which limits suitability as inhibitor of T-type voltage-gated Ca(2+) channels.
ACS chemical neuroscience (2014-12-02)
Katherine R Brimblecombe, Stephanie J Cragg
ABSTRACT
Neuronal T-type voltage-gated Ca(2+) channels are reported to have physiological roles that include regulation of burst firing, Ca(2+) oscillations, and neurotransmitter release. These roles are often exposed experimentally by blocking T-type channels with micromolar Ni(2+). We used Ni(2+) to explore the role of axonal T-type channels in dopamine (DA) release in mouse striatum, but identified significant off-target effects on DA uptake. Ni(2+) (100 μM) reversibly increased electrically evoked DA release and markedly extended its extracellular lifetime, detected using fast-scan cyclic voltammetry. Prior inhibition of the DA transporter (DAT) by cocaine (5 μM) occluded the facilitatory action of Ni(2+) on DA release and conversely, allowed Ni(2+) to inhibit release, presumably through T-channel inhibition. Ni(2+) further prolonged the timecourse of DA clearance suggesting further inhibition of DA uptake. In summary, Ni(2+) has major effects on DA transmission besides those due to T-channels that likely involve inhibition of the DAT.
Sigma-Aldrich
Cocaine free base
Sigma-Aldrich
Nickel(II) chloride, 98%
Sigma-Aldrich
Nickel(II) chloride, anhydrous, powder, 99.99% trace metals basis