Merck
  • Embedding islet in a liquid scaffold increases islet viability and function.
Embedding islet in a liquid scaffold increases islet viability and function.
Canadian journal of diabetes (2013-09-28)
Azadeh Hosseini-Tabatabaei, Reza Baradar Jalili, Ryan Hartwell, Sanam Salimi, Ruhangiz T Kilani, Aziz Ghahary
ABSTRACT
Islet transplantation is a promising strategy to restore efficient insulin regulation in type 1 diabetes mellitus patients. However, shortage of islet donors, poor islet survival and toxicity of immunosuppressants often reduce the graft functional lifetime. We previously showed that a fibroblast populated-collagen matrix (CM) significantly improved engrafted islet viability/function. However, this composite was prone to gradual biodegradation and contraction. Moreover, to avoid use of systemic immunosuppressants, we proposed the use of a local immunosuppressive enzyme, indoleamine-2,3-dioxygenase (IDO). We developed a novel bioengineered crosslinked CM (CCM) to provide optimal matrix biomimetic. Viability and insulin secretory function of islets embedded within fibroblast populated CCM (FP-CCM) was evaluated in vitro and in vivo. IDO expression was transduced in fibroblasts by a lentiviral vector carrying IDO gene and islet viability was evaluated in the presence and absence of IDO producing cells. Islet survival/function markedly improved within FP-CCM. Furthermore, our data shows that local lentiviral induction of IDO delivered by FP-CCM is nontoxic to the embedded islets. This promising finding offers a new approach to improving islet transplant outcome.
Product Description
Amphotericin B, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
Streptozocin, ≥75% α-anomer basis, ≥98% (HPLC), powder