Merck
The senescent hepatocyte gene signature in chronic liver disease.
Experimental gerontology (2014-09-23)
Aloysious Aravinthan, Nicholas Shannon, Judith Heaney, Matthew Hoare, Aileen Marshall, Graeme J M Alexander
ABSTRACT
Hepatocyte senescence is associated closely with fibrosis stage and an adverse outcome in chronic liver disease, but it is uncertain whether there is a causal relation with clinical manifestations of chronic liver disease, which was the subject of this study of the senescent hepatocyte gene signature. Senescence was induced in HepG2 cells using sub-lethal concentrations of H2O2. Gene expression of control and senescent HepG2 cells were studied. Comparison was made with patients with cirrhosis and three public microarray datasets. H2O2-treated HepG2 cells demonstrated characteristic cellular senescence. There was differential expression of 354 genes in senescence. Up-regulated genes in HepG2 senescence were also up regulated in patients with cirrhosis. The senescent hepatocyte gene signature distinguished liver disease from normal by unsupervised clustering in the public chronic liver disease microarray datasets, with enrichment of the senescence gene signature in all three datasets. The senescent hepatocyte gene signature included changes in cell cycle regulation, morphology, inflammation, signal transduction, metabolism and stellate cell activation, which alongside impaired synthetic function in senescence in vitro were consistent with manifestations of clinical liver disease, suggesting a close relation between hepatocyte senescence and manifestations of chronic liver disease including fibrosis and impaired synthetic function.
Sigma-Aldrich
Phosphorus, red, ≥99.99% trace metals basis
Sigma-Aldrich
Phosphorus, red, ≥97.0%