Merck
  • Synthesis and activity on rat aorta rings and rat pancreatic beta-cells of ring-opened analogues of benzopyran-type potassium channel activators.
Synthesis and activity on rat aorta rings and rat pancreatic beta-cells of ring-opened analogues of benzopyran-type potassium channel activators.
Bioorganic & medicinal chemistry (2008-05-16)
Smail Khelili, Xavier Florence, Mourad Bouhadja, Samia Abdelaziz, Nadia Mechouch, Yekhlef Mohamed, Pascal de Tullio, Philippe Lebrun, Bernard Pirotte
ABSTRACT
Ring-opened analogues of dihydrobenzopyran potassium channel openers (PCOs) were prepared and evaluated as putative PCOs on rat aorta rings (myorelaxant effect) and rat pancreatic beta-cells (inhibition of insulin secretion). These derivatives are characterized by the presence of a sulfonylurea, a urea or an amide function. Some compounds bearing an arylurea moiety provoked vasorelaxant effects and a marked inhibition of insulin release. Derivatives bearing a sulfonylurea or an amide function were, however, poorly active on both tissues. Structure-activity relationships and apparent tissue selectivity are discussed.
Sigma-Aldrich
3-Cyanophenyl isocyanate, 97%
Pricing and availability is not currently available.