Merck
  • Worker illness related to newly marketed pesticides--Douglas County, Washington, 2014.
Worker illness related to newly marketed pesticides--Douglas County, Washington, 2014.
MMWR. Morbidity and mortality weekly report (2015-01-23)
Geoffrey M Calvert, Luis Rodriguez, Joanne Bonnar Prado
ABSTRACT
On April 10, 2014 the Washington State Department of Agriculture (WSDA) was notified by a local newspaper of a suspected pesticide poisoning incident in Douglas County involving pesticides not previously reported in the published literature to be associated with human illness. On that same day, WSDA notified the Washington State Department of Health, which investigated this incident by conducting a site visit, reviewing medical and applicator records, and interviewing affected farmworkers, pesticide applicators, and the farmworkers' employer. In addition, on April 11, WSDA collected swab, foliage, and clothing samples and tested them for residues of pyridaben, novaluron, and triflumizole. In this incident, all 20 farmworkers working in a cherry orchard became ill from off-target drift of a pesticide mixture that was being applied to a neighboring pear orchard. Sixteen sought medical treatment for neurologic, gastrointestinal, ocular, and respiratory symptoms. This event highlights the need for greater efforts to prevent off-target drift exposures and promote awareness about the toxicity of some recently marketed pesticides. Incidents such as this could be prevented if farm managers planning pesticide applications notify their neighbors of their plans.
Product Description
Supelco
Pyridaben, PESTANAL®, analytical standard
Supelco
Triflumizole, PESTANAL®, analytical standard