Merck
  • Identification of insecticidal constituents of the essential oils of Dahlia pinnata Cav. against Sitophilus zeamais and Sitophilus oryzae.
Identification of insecticidal constituents of the essential oils of Dahlia pinnata Cav. against Sitophilus zeamais and Sitophilus oryzae.
Natural product research (2015-01-08)
Da-Cheng Wang, Da-Ren Qiu, Li-Na Shi, Hong-Yu Pan, Ya-Wei Li, Jin-Zhu Sun, Ying-Jie Xue, Dong-Sheng Wei, Xiang Li, Ya-Mei Zhang, Jian-Chun Qin
ABSTRACT
The aim of this research was to determine the chemical composition of the essential oils of Dahlia pinnata, their insecticidal activity against Sitophilus zeamais and Sitophilusoryzae and to isolate insecticidal constituents. Based on bioactivity-guided fractionation, active constituents were isolated and identified as D-limonene, 4-terpineol and α-terpineol. Essential oils and active compounds tested exhibited contact toxicity, with LD50 values ranging from 132.48 to 828.79 μg/cm(2) against S. zeamais and S. oryzae. Essential oils possessed fumigant toxicity against S. zeamais and S. oryzae with LC50 from 14.10 to 73.46 mg/L. d-Limonene (LC50 = 4.55 and 7.92 mg/L) showed stronger fumigant toxicity against target insects. 4-Terpineol (88 ± 8%) and d-limonene (87 ± 5%) showed the strongest repellency against S. zeamais and S. oryzae, respectively. The results indicate that essential oils and insecticidal constituents have potential for development into natural fumigants, insecticides or repellents for control of the stored-product insect pests.
Sigma-Aldrich
Terpineol, mixture of isomers, 96%, FG
Pricing and availability is not currently available.