Merck
  • Autochthonous blastomycosis of the adrenal: first case report from Asia.
Autochthonous blastomycosis of the adrenal: first case report from Asia.
The American journal of tropical medicine and hygiene (2014-02-05)
Anil Kumar, Sreekala Sreehari, Kandan Velayudhan, Lalitha Biswas, Rachana Babu, Shabeer Ahmed, Neelakanta Sharma, Vasanth P Kurupath, Annie Jojo, Kavitha R Dinesh, Shamsul Karim, Raja Biswas
ABSTRACT
Systemic endemic mycoses, such as blastomycosis, are rare in Asia and have been reported as health risks among travelers who visit or reside in an endemic area. Adrenal involvement is rarely seen in blastomycosis and has never been reported from Asia. We report the first case of blastomycosis with bilateral involvement of the adrenals in a diabetic patient residing in the state of Arunachal Pradesh, India.
Product Description
Itraconazole, European Pharmacopoeia (EP) Reference Standard
Itraconazole for system suitability, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
Itraconazole, ≥98% (HPLC)
Supelco
Itraconazole solution, 2.0 mg/mL (Methanol with 1% 1M HCl), ampule of 1 mL, certified reference material, Cerilliant®