Merck
  • [Research progress of alternative production approaches of salidroside].
[Research progress of alternative production approaches of salidroside].
Zhongguo Zhong yao za zhi = Zhongguo zhongyao zazhi = China journal of Chinese materia medica (2014-02-06)
Xiu-Wen Wu, Yu-Shuai Peng, Ru-Feng Wang
ABSTRACT
Salidroside, one of the active components of Rhodiola plants, is a phenolic glycoside with significant biological activities. The investigation and development of alternative production approaches of salidroside is of high academic and application values due to the limited resource of Rhodiola plants, and from which the low yield of salidroside. This review summarized the research progress and perspective of the alternative production approaches of salidroside including both chemosynthetic and biosynthetic methods and pathways.
Sigma-Aldrich
Salidroside, ≥95% (LC/MS-ELSD)
Supelco
Salidroside, analytical standard