Merck
  • Characterization of ferrioxamine E as the principal siderophore of Erwinia herbicola (Enterobacter agglomerans).
Characterization of ferrioxamine E as the principal siderophore of Erwinia herbicola (Enterobacter agglomerans).
Biology of metals (1988-01-01)
I Berner, S Konetschny-Rapp, G Jung, G Winkelmann
ABSTRACT
Several strains of the enterobacterial group Erwinia herbicola (Enterobacter agglomerans) were screened for siderophore production. After 3 days of growth in a low-iron medium, all strains studied produced hydroxamate siderophores. The retention values of the main siderophore during thin-layer chromatography on silica gel plates and on HPLC reversed-phase columns were identical with those of an authentic sample of ferrioxamine E (norcardamine). Gas-chromatographic analysis of the HI hydrolyzate yielded succinic acid and 1,5-diaminopentane in equimolar amounts; fast-atom-bombardment (FAB) mass spectroscopy showed a molecular mass of 653 Da. Iron from 55Fe-labelled ferrioxamine E was well taken up by iron-starved cells of E. herbicola (Km = 0.1 microM, Vmax = 8 pmol mg-1 min-1). However, besides ferrioxamine E (100%), several exogenous siderophores such as enterobactin (94.5%), ferric citrate (78.5%), coprogen (63.5%) and ferrichrome (17.5%) served as siderophores, suggesting the presence of multiple siderophore receptors in the outer membrane of E. herbicola.
Product Description
Millipore
Ferrioxamine E from Streptomyces antibioticus, suitable for microbiology, ≥95%
Pricing and availability is not currently available.